2-Methylimidazo[1,2-a]pyridine 95%

Supplier: Thermo Scientific Chemicals

Warning

H35114-03 H35114-03
AAH35114-03EA 176.73 USD
CAAAH35114-03 AAH35114-03
2-Methylimidazo[1,2-a]pyridine 95%
2-Methylimidazo[1,2-a]pyridine
Formula: C₈H₈N₂
MW: 132.17 g/mol
Boiling Pt: 148 °C/20 mm
Melting Pt: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD02251330
CAS Number: 934-37-2

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR