3-Chloroimidazo[1,2-a]pyridine-6-carboxylic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H35089-MD
H35089-03
H35089-MD
H35089-03
AAH35089-MDEA
85.79
USD
CAAAH35089-03
CAAAH35089-MD
AAH35089-MD
AAH35089-03
3-Chloroimidazo[1,2-a]pyridine-6-carboxylic acid 95%
3-Chloroimidazo[1,2-a]pyridine-6-carboxylic acid
Formula:
C₈H₅ClN₂O₂ Melting Pt: ca 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD07021510 CAS Number: 900019-39-8 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...