Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H35022-03
H35022-MD
H35022-MD
H35022-03
AAH35022-MDEA
97.23
USD
CAAAH35022-MD
AAH35022-MD
AAH35022-03
CAAAH35022-03
Ethyl-3-bromoimidazo[1,2-a]pyridine-6-carboxylate 95%
Ethyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate
Formula:
C₁₀H₉BrN₂O₂ MW: 269.10 Melting Pt: 95…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD12827886 CAS Number: 1215504-30-5 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...