2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide 97%

Danger

Synonyms: 2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide

H34482-MD H34482-MD H34482-03 H34482-03
AAH34482-MDEA 67.49 USD
AAH34482-MD CAAAH34482-MD AAH34482-03 CAAAH34482-03
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide 97%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide
Formula: C₉H₇BrF₄
MW: 271.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687198
CAS Number: 1323966-23-9
UN: 3265
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR