4-Chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine 96%
|
Warning
|
H34471-06
H34471-MD
H34471-03
H34471-03
H34471-06
H34471-MD
AAH34471-03EA
67.08
USD
CAAAH34471-06
CAAAH34471-MD
AAH34471-03
CAAAH34471-03
AAH34471-06
AAH34471-MD
4-Chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine 96%
4-Chloro-2,7-dimethyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidine
Formula:
C₁₂H₁₃ClN₂S Melting Pt: 91…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD03085777 CAS Number: 610274-01-6 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...