2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride 97%

Danger

Synonyms: 2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride

H34468-03 H34468-MD H34468-MD H34468-03
AAH34468-MDEA 205.35 USD
CAAAH34468-MD AAH34468-MD AAH34468-03 CAAAH34468-03
2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride 97%
2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Formula: C₇H₅ClF₃NO₂S
MW: 259.64 g/mol
Melting Pt: 57…59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14583150
CAS Number: 1257535-05-9
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR