Ethyl-2-amino-4-(4-chlorophenyl)thiophene-3-carboxylate 96%

Warning

H34427-03 H34427-09 H34427-03 H34427-09
AAH34427-09EA 216.45 USD
CAAAH34427-03 CAAAH34427-09 AAH34427-09 AAH34427-03
Ethyl-2-amino-4-(4-chlorophenyl)thiophene-3-carboxylate 96%
Ethyl-2-amino-4-(4-chlorophenyl)thiophene-3-carboxylate
Formula: C₁₃H₁₂ClNO₂S
MW: 281.754 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD01038372
CAS Number: 65234-09-5

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR