3-Chloro-4-(difluoromethyl)pyridine 97%
|
Warning
|
H34426-03
H34426-MD
H34426-MD
H34426-03
AAH34426-MDEA
147.63
USD
CAAAH34426-03
CAAAH34426-MD
AAH34426-MD
AAH34426-03
3-Chloro-4-(difluoromethyl)pyridine 97%
3-Chloro-4-(difluoromethyl)pyridine
Formula:
C₆H₄ClF₂N Storage Temperature: Ambient |
MDL Number:
MFCD22124921 CAS Number: 1374659-44-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...