3'-methylbiphenyl-4-sulfonyl chloride 96%

Danger

H34394-MD H34394-06 H34394-03 H34394-06 H34394-MD
AAH34394-MDEA 39.65 USD
CAAAH34394-03 AAH34394-MD AAH34394-06 CAAAH34394-06 CAAAH34394-MD
3'-methylbiphenyl-4-sulfonyl chloride 96%
3'-methylbiphenyl-4-sulfonyl chloride
Formula: C₁₃H₁₁ClO₂S
MW: 266.739 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD06409269
CAS Number: 186551-47-3
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR