4'-Fluorobiphenyl-4-carboxaldehyde 96%
|
Warning
|
H34387-MD
H34387-03
H34387-03
H34387-MD
AAH34387-MDEA
33.25
USD
AAH34387-MD
CAAAH34387-03
AAH34387-03
CAAAH34387-MD
4'-Fluorobiphenyl-4-carboxaldehyde 96%
4'-Fluorobiphenyl-4-carboxaldehyde
Formula:
C₁₃H₉FO MW: 200.212 g/mol Boiling Pt: 324 °C Melting Pt: 93…97 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01631912 CAS Number: 60992-98-5 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...