1-Phenylcyclobutanecarboxylic acid 97%
|
Warning
|
H34382-MD
H34382-MD
H34382-03
H34382-03
AAH34382-03EA
216.45
USD
CAAAH34382-03
AAH34382-03
AAH34382-MD
CAAAH34382-MD
1-Phenylcyclobutanecarboxylic acid 97%
1-Phenylcyclobutanecarboxylic acid
Formula:
C₁₀H₁₀O₂ MW: 176.22 g/mol Melting Pt: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019262 CAS Number: 37828-19-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...