3-Methoxy-4-(trifluoromethyl)benzonitrile 97%
|
Warning
|
H34366-06
H34366-06
H34366-03
H34366-03
AAH34366-06EA
426.24
USD
AAH34366-06
AAH34366-03
CAAAH34366-03
CAAAH34366-06
3-Methoxy-4-(trifluoromethyl)benzonitrile 97%
3-Methoxy-4-(trifluoromethyl)benzonitrile
Formula:
C₉H₆F₃NO Melting Pt: 52…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD13185685 CAS Number: 447-93-8 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...