4-Fluoro-2-(trifluoromethoxy)benzyl alcohol 97%
|
Warning
|
H34362-03
H34362-MD
H34362-03
H34362-MD
AAH34362-MDEA
28.09
USD
AAH34362-MD
CAAAH34362-MD
AAH34362-03
CAAAH34362-03
4-Fluoro-2-(trifluoromethoxy)benzyl alcohol 97%
4-Fluoro-2-(trifluoromethoxy)benzyl alcohol
Formula:
C₈H₆F₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD19687174 CAS Number: 1242252-68-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...