2,5-Dichloro-3-(difluoromethyl)pyridine 95%
|
Warning
|
H34359-MD
H34359-03
H34359-03
H34359-MD
AAH34359-03EA
307.47
USD
CAAAH34359-MD
AAH34359-03
AAH34359-MD
CAAAH34359-03
2,5-Dichloro-3-(difluoromethyl)pyridine 95%
2,5-Dichloro-3-(difluoromethyl)pyridine
Formula:
C₆H₃Cl₂F₂N Boiling Pt: 69 °C/7 mm Storage Temperature: Ambient |
MDL Number:
MFCD18384828 CAS Number: 71701-93-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...