4-Chloro-3-(trifluoromethoxy)benzylamine 97%
|
Danger
|
H34356-MD
H34356-MD
H34356-03
H34356-03
AAH34356-03EA
76.22
USD
CAAAH34356-03
AAH34356-03
AAH34356-MD
CAAAH34356-MD
4-Chloro-3-(trifluoromethoxy)benzylamine 97%
4-Chloro-3-(trifluoromethoxy)benzylamine
Formula:
C₈H₇ClF₃NO Storage Temperature: Ambient |
MDL Number:
MFCD18393827 CAS Number: 916210-69-0 UN: 2735 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...