4-Ethoxy-3-(trifluoromethyl)benzyl alcohol 97%
|
Warning
|
Synonyms:
4-ETHOXY-3-(TRIFLUOROMETHYL)BENZYL ALCOHOL
H34346-06
H34346-03
H34346-03
H34346-06
AAH34346-06EA
342.99
USD
CAAAH34346-06
AAH34346-06
AAH34346-03
CAAAH34346-03
4-Ethoxy-3-(trifluoromethyl)benzyl alcohol 97%
4-Ethoxy-3-(trifluoromethyl)benzyl alcohol
Formula:
C₁₀H₁₀F₃NO₂ MW: 233.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13196581 CAS Number: 1206593-33-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...