3-Chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde 97%
|
Warning
|
H34338-03
H34338-MD
H34338-MD
H34338-03
AAH34338-MDEA
38.75
USD
AAH34338-MD
AAH34338-03
CAAAH34338-03
CAAAH34338-MD
3-Chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde 97%
3-Chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde
Formula:
C₉H₈ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22201016 CAS Number: 1373920-69-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...