4-n-Butoxy-2,6-difluorophenol 97%

Warning

Synonyms: 4-Butoxy-2,6-difluorophenol

H34337-03 H34337-MD H34337-MD H34337-03
AAH34337-MDEA 61.22 USD
AAH34337-MD CAAAH34337-MD AAH34337-03 CAAAH34337-03
4-n-Butoxy-2,6-difluorophenol 97%
4-n-Butoxy-2,6-difluorophenol
Formula: C₁₀H₁₂F₂O₂
MW: 202.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201025
CAS Number: 1373920-65-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR