2-Methyl-4-trifluoromethoxy-DL-phenylalanine 97%

Synonyms: 2-Methyl-4-(trifluoromethoxy)-DL-phenylalanine

H34333-MD H34333-03 H34333-MD H34333-03
AAH34333-03EA 198.69 USD
CAAAH34333-03 AAH34333-03 CAAAH34333-MD AAH34333-MD
2-Methyl-4-trifluoromethoxy-DL-phenylalanine 97%
2-Methyl-4-trifluoromethoxy-DL-phenylalanine
Formula: C₁₁H₁₂F₃NO₃
MW: 263.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201062
CAS Number: 1706428-65-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR