3'-Methoxybiphenyl-4-carboxaldehyde 96%

Warning

H34312-06 H34312-MD H34312-03 H34312-03 H34312-MD H34312-06
AAH34312-03EA 98.45 USD
AAH34312-03 CAAAH34312-MD CAAAH34312-06 CAAAH34312-03 AAH34312-MD AAH34312-06
3'-Methoxybiphenyl-4-carboxaldehyde 96%
3'-Methoxybiphenyl-4-carboxaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.248 g/mol
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03424619
CAS Number: 209863-09-2

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR