3'-Chloro-5'-(pentafluorothio)acetophenone 97%

Warning

H34278-03 H34278-MD H34278-03 H34278-MD
AAH34278-MDEA 94.39 USD
AAH34278-MD CAAAH34278-MD CAAAH34278-03 AAH34278-03
3'-Chloro-5'-(pentafluorothio)acetophenone 97%
3'-Chloro-5'-(pentafluorothio)acetophenone
Formula: C₈H₆ClF₅OS
MW: 280.64
Storage Temperature: Ambient
MDL Number: MFCD16652455
CAS Number: 1180486-96-7

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR