4-Chloro-2-fluoro-3-methoxyaniline 97%

Warning

Synonyms: 4-chloro-2-fluoro-3-methoxyaniline

H34255-03 H34255-03 H34255-MD H34255-MD
AAH34255-MDEA 92.59 USD
AAH34255-MD CAAAH34255-03 AAH34255-03 CAAAH34255-MD
4-Chloro-2-fluoro-3-methoxyaniline 97%
4-Chloro-2-fluoro-3-methoxyaniline
Formula: C₇H₇ClFNO
MW: 175.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687207
CAS Number: 1323966-39-7
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR