N-(6-Chloro-4-pyrimidinyl)-N-(2-furylmethyl)amine 96%

Warning

Synonyms: 6-Chloro-4-(furfurylamino)pyrimidine

H34241-ME H34241-ME
AAH34241-MEEA 138.4 USD
AAH34241-ME CAAAH34241-ME
N-(6-Chloro-4-pyrimidinyl)-N-(2-furylmethyl)amine 96%
N-(6-Chloro-4-pyrimidinyl)-N-(2-furylmethyl)amine
Formula: C₉H₈ClN₃O
MW: 209.633 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD11655644
CAS Number: 99846-86-3

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR