4-Chloro-2-trifluoromethoxy-DL-phenylalanine 97%

Synonyms: 4-Chloro-2-trifluoromethoxy-DL-phenylalanine

H34226-03 H34226-MD H34226-03 H34226-MD
AAH34226-03EA 275.28 USD
AAH34226-03 CAAAH34226-MD AAH34226-MD CAAAH34226-03
4-Chloro-2-trifluoromethoxy-DL-phenylalanine 97%
4-Chloro-2-trifluoromethoxy-DL-phenylalanine
Formula: C₁₀H₉ClF₃NO₃
MW: 283.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687214
CAS Number: 1435806-19-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR