2-Amino-4-(2-bromophenyl)pyrimidine 95%
|
Warning
|
H34212-03
H34212-06
H34212-06
H34212-03
AAH34212-03EA
67.08
USD
CAAAH34212-03
CAAAH34212-06
AAH34212-03
AAH34212-06
2-Amino-4-(2-bromophenyl)pyrimidine 95%
2-Amino-4-(2-bromophenyl)pyrimidine
Formula:
C₁₀H₈BrN₃ Melting Pt: 167…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD07381905 CAS Number: 99073-95-7 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...