(3,5-Difluorophenyl)methanesulfonyl chloride 95%

Danger

Synonyms: 3,5-Difluoro-α-toluenesulphonyl chloride

H34210-MD H34210-03 H34210-MD H34210-03
AAH34210-03EA 188.81 USD
CAAAH34210-MD AAH34210-03 AAH34210-MD CAAAH34210-03
(3,5-Difluorophenyl)methanesulfonyl chloride 95%
(3,5-Difluorophenyl)methanesulphonyl chloride
Formula: C₇H₅ClF₂O₂S
MW: 226.63 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09743349
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR