4-Fluoro-2-(trifluoromethoxy)benzyl bromide 97%
|
Danger
|
Synonyms:
1-(Bromomethyl)-4-fluoro-2-(trifluoromethoxy)benzene
H34202-MD
H34202-MD
H34202-03
H34202-03
AAH34202-03EA
216.45
USD
CAAAH34202-MD
AAH34202-03
AAH34202-MD
CAAAH34202-03
4-Fluoro-2-(trifluoromethoxy)benzyl bromide 97%
4-Fluoro-2-(trifluoromethoxy)benzyl bromide
Formula:
C₈H₅BrF₄O MW: 273.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687175 CAS Number: 1323966-22-8 UN: 3265 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...