4-(4-Trifluoromethyl-2-pyrimidinyloxy)benzenesulfonyl chloride 95%

Danger

H34198-MD H34198-03 H34198-03 H34198-MD
AAH34198-03EA 303.03 USD
AAH34198-03 CAAAH34198-MD AAH34198-MD CAAAH34198-03
4-(4-Trifluoromethyl-2-pyrimidinyloxy)benzenesulfonyl chloride 95%
4-(4-Trifluoromethyl-2-pyrimidinyloxy)benzenesulfonyl chloride
Formula: C₁₁H₆ClF₃N₂O₃S
MW: 338.685 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD11109532
CAS Number: 1160058-86-5
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR