3,5-Difluoro-2-methylbenzonitrile 97%
|
Warning
|
H34195-MD
H34195-03
H34195-MD
H34195-03
AAH34195-03EA
236.43
USD
AAH34195-03
CAAAH34195-MD
CAAAH34195-03
AAH34195-MD
3,5-Difluoro-2-methylbenzonitrile 97%
3,5-Difluoro-2-methylbenzonitrile
Formula:
C₈H₅F₂N Melting Pt: 55…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD09996934 CAS Number: 1003708-74-4 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...