2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde 97%

Warning

Synonyms: 2-fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde

H34174-06 H34174-03 H34174-06 H34174-03
AAH34174-03EA 100.56 USD
CAAAH34174-03 CAAAH34174-06 AAH34174-03 AAH34174-06
2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde
Formula: C₉H₆F₄O
MW: 206.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687179
CAS Number: 1323966-40-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR