2-Methyl-5-(trifluoromethoxy)benzonitrile 97%
|
Warning
|
H34142-03
H34142-03
H34142-06
H34142-06
AAH34142-06EA
369.63
USD
CAAAH34142-03
CAAAH34142-06
AAH34142-06
AAH34142-03
2-Methyl-5-(trifluoromethoxy)benzonitrile 97%
2-Methyl-5-(trifluoromethoxy)benzonitrile
Formula:
C₉H₆F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD18399750 CAS Number: 1261679-13-3 UN: 3276 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...