4-Chloro-2-(trifluoromethoxy)benzylamine 97%

Danger

H34140-MD H34140-MD H34140-03 H34140-03
AAH34140-MDEA 81.14 USD
CAAAH34140-03 AAH34140-MD AAH34140-03 CAAAH34140-MD
4-Chloro-2-(trifluoromethoxy)benzylamine 97%
4-Chloro-2-(trifluoromethoxy)benzylamine
Formula: C₈H₇ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD18393826
CAS Number: 1261605-72-4
UN: 3267
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR