4-Chloro-2-(trifluoromethoxy)aniline 97%

Warning

Synonyms: 4-Chloro-2-(trifluoromethoxy)phenylamine

H34133-MD H34133-03 H34133-03 H34133-MD
AAH34133-03EA 158.73 USD
AAH34133-03 AAH34133-MD CAAAH34133-03 CAAAH34133-MD
4-Chloro-2-(trifluoromethoxy)aniline 97%
4-Chloro-2-(trifluoromethoxy)aniline
Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
Boiling Pt: 82 °C/15 mm
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00153102
CAS Number: 175205-77-3
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR