1H-Indazole-4-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals

Warning

H34115-03 H34115-MD H34115-03 H34115-06 H34115-MD H34115-06
AAH34115-MDEA 28.49 USD
CAAAH34115-03 AAH34115-MD AAH34115-06 AAH34115-03 CAAAH34115-06 CAAAH34115-MD
1H-Indazole-4-carboxylic acid 97%
1H-Indazole-4-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: ca 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03840644
CAS Number: 677306-38-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR