2-Methyl-5-(trifluoromethoxy)benzoic acid 97%

Warning

H34102-03 H34102-06 H34102-03 H34102-06
AAH34102-06EA 271.95 USD
AAH34102-06 AAH34102-03 CAAAH34102-03 CAAAH34102-06
2-Methyl-5-(trifluoromethoxy)benzoic acid 97%
2-Methyl-5-(trifluoromethoxy)benzoic acid
Formula: C₉H₇F₃O₃
MW: 220.147 g/mol
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD18399742
CAS Number: 195622-41-4

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR