4-Chloro-2-(trifluoromethoxy)benzyl alcohol 97%
|
Warning
|
H34078-MD
H34078-03
H34078-MD
H34078-03
AAH34078-03EA
158.73
USD
AAH34078-03
CAAAH34078-MD
AAH34078-MD
CAAAH34078-03
4-Chloro-2-(trifluoromethoxy)benzyl alcohol 97%
4-Chloro-2-(trifluoromethoxy)benzyl alcohol
Formula:
C₈H₆ClF₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18393819 CAS Number: 1261673-90-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...