4-Chloro-3-ethoxy-2-fluorophenylacetic acid 97%

Warning

Synonyms: 4-Chloro-3-ethoxy-2-fluorophenylacetic acid

H34073-MD H34073-03 H34073-MD H34073-03
AAH34073-03EA 316.35 USD
CAAAH34073-MD AAH34073-03 CAAAH34073-03 AAH34073-MD
4-Chloro-3-ethoxy-2-fluorophenylacetic acid 97%
4-Chloro-3-ethoxy-2-fluorophenylacetic acid
Formula: C₁₀H₁₀ClFO₃
MW: 232.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687188
CAS Number: 1323955-63-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR