3,3',5-Triiodo-L-thyronine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-2-Amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoic acid
H34068-03
H34068-03
H34068-06
H34068-MD
H34068-06
H34068-MD
AAH34068-03EA
251.99
USD
AAH34068-03
AAH34068-06
AAH34068-MD
CAAAH34068-MD
CAAAH34068-06
CAAAH34068-03
3,3',5-Triiodo-L-thyronine 95%
3,3',5-Triiodo-L-thyronine
Formula:
C₁₅H₁₂I₃NO₄ MW: 650.98 g/mol Melting Pt: 234…238 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002593 CAS Number: 6893-02-3 EINECS: 229-999-3 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...