2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline 97%
|
Warning
|
Synonyms:
2-fluoro-5-methoxy-3-(trifluoromethyl)aniline
H34066-06
H34066-03
H34066-06
H34066-03
AAH34066-03EA
60.98
USD
AAH34066-03
CAAAH34066-06
AAH34066-06
CAAAH34066-03
2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline
Formula:
C₈H₇F₄NO MW: 209.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201043 CAS Number: 1262198-05-9 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...