4-n-Butoxy-2,6-difluorobenzamide 97%
|
Warning
|
Synonyms:
4-Butoxy-2,6-difluorobenzamide
H34027-MD
H34027-MD
AAH34027-MDEA
113.22
USD
AAH34027-MD
CAAAH34027-MD
4-n-Butoxy-2,6-difluorobenzamide 97%
4-n-Butoxy-2,6-difluorobenzamide
Formula:
C₁₁H₁₃F₂NO₂ MW: 229.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201024 CAS Number: 1373920-96-7 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...