4-Methoxy-3-(trifluoromethoxy)cinnamic acid 97%

Warning

Synonyms: 4-Methoxy-3-(trifluoromethoxy)cinnamic acid

H33982-MD H33982-MD H33982-03 H33982-03
AAH33982-03EA 198.69 USD
CAAAH33982-03 AAH33982-03 AAH33982-MD CAAAH33982-MD
4-Methoxy-3-(trifluoromethoxy)cinnamic acid 97%
4-Methoxy-3-(trifluoromethoxy)cinnamic acid
Formula: C₁₁H₉F₃O₄
MW: 262.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398503
CAS Number: 1262018-46-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR