3,3'-Di-n-pentylthiadicarbocyanine iodide 96%

Warning

H33978-ME H33978-ME
AAH33978-MEEA 150.96 USD
AAH33978-ME CAAAH33978-ME
3,3'-Di-n-pentylthiadicarbocyanine iodide 96%
3,3'-Di-n-pentylthiadicarbocyanine iodide
Formula: C₂₉H₃₅IN₂S₂
MW: 602.637 g/mol
Melting Pt: ca 212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07433877
CAS Number: 53213-96-0

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR