2-Fluoro-5-methyl-4-(trifluoromethyl)benzamide 97%

Warning

Synonyms: 2-fluoro-5-methyl-4-(trifluoromethyl)benzamide

H33960-03 H33960-MD H33960-MD H33960-03
AAH33960-03EA 198.69 USD
CAAAH33960-MD AAH33960-03 AAH33960-MD CAAAH33960-03
2-Fluoro-5-methyl-4-(trifluoromethyl)benzamide 97%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzamide
Formula: C₉H₇F₄NO
MW: 221.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687194
CAS Number: 1323966-43-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR