4-Methoxy-3-(trifluoromethoxy)benzyl bromide 97%
|
Danger
|
H33950-03
H33950-MD
H33950-03
H33950-MD
AAH33950-03EA
198.69
USD
AAH33950-03
CAAAH33950-MD
CAAAH33950-03
AAH33950-MD
4-Methoxy-3-(trifluoromethoxy)benzyl bromide 97%
4-Methoxy-3-(trifluoromethoxy)benzyl bromide
Formula:
C₉H₈BrF₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18398486 CAS Number: 853771-92-3 UN: 3265 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...