4-Chloro-2-fluoro-3-methoxyphenylacetonitrile 97%
|
Warning
|
Synonyms:
4-Chloro-2-fluoro-3-methoxyphenylacetonitrile
H33939-MD
H33939-03
H33939-MD
H33939-03
AAH33939-MDEA
92.59
USD
AAH33939-MD
AAH33939-03
CAAAH33939-03
CAAAH33939-MD
4-Chloro-2-fluoro-3-methoxyphenylacetonitrile 97%
4-Chloro-2-fluoro-3-methoxyphenylacetonitrile
Formula:
C₉H₇ClFNO MW: 199.61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687183 CAS Number: 1323966-29-5 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...