2-Methyl-5-(trifluoromethoxy)benzyl bromide 96%
|
Danger
|
H33930-06
H33930-03
H33930-03
H33930-06
AAH33930-03EA
133.2
USD
CAAAH33930-06
CAAAH33930-03
AAH33930-03
AAH33930-06
2-Methyl-5-(trifluoromethoxy)benzyl bromide 96%
2-Methyl-5-(trifluoromethoxy)benzyl bromide
Formula:
C₉H₈BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD18399774 CAS Number: 1261573-60-7 UN: 3265 ADR: "8",III |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...