3-(Benzyloxy)cyclobutane-1,1-dicarboxylic acid 95%

Warning

H33919-03 H33919-MD H33919-MD H33919-03
AAH33919-03EA 275.28 USD
AAH33919-03 AAH33919-MD CAAAH33919-MD CAAAH33919-03
3-(Benzyloxy)cyclobutane-1,1-dicarboxylic acid 95%
3-(Benzyloxy)cyclobutane-1,1-dicarboxylic acid
Formula: C₁₃H₁₄O₅
MW: 250.25 g/mol
Melting Pt: ca 164 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD11559004
CAS Number: 84182-46-7

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR