5-Bromo-3-iodo-6-methylpyridin-2-amine 95%

Warning

Synonyms: 2-Amino-5-bromo-3-iodo-6-methylpyridine

H33888-MD H33888-03 H33888-MD H33888-03
AAH33888-MDEA 33.01 USD
CAAAH33888-MD CAAAH33888-03 AAH33888-MD AAH33888-03
5-Bromo-3-iodo-6-methylpyridin-2-amine 95%
5-Bromo-3-iodo-6-methylpyridin-2-amine
Formula: C₆H₆BrIN₂
MW: 312.936 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD09880093
CAS Number: 958357-86-3

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR