3-Methoxy-4-(trifluoromethyl)benzyl bromide 97%
|
Danger
|
H33880-03
H33880-03
H33880-MD
H33880-MD
AAH33880-03EA
152.52
USD
AAH33880-03
AAH33880-MD
CAAAH33880-03
CAAAH33880-MD
3-Methoxy-4-(trifluoromethyl)benzyl bromide 97%
3-Methoxy-4-(trifluoromethyl)benzyl bromide
Formula:
C₉H₈BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD18398596 CAS Number: 853367-87-0 UN: 3261 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...