4-Chloro-3-ethoxy-2-fluorobenzoic acid 97%

Warning

Synonyms: 4-chloro-3-ethoxy-2-fluorobenzoic acid

H33868-03 H33868-06 H33868-03 H33868-06
AAH33868-06EA 496.17 USD
AAH33868-06 CAAAH33868-03 CAAAH33868-06 AAH33868-03
4-Chloro-3-ethoxy-2-fluorobenzoic acid 97%
4-Chloro-3-ethoxy-2-fluorobenzoic acid
Formula: C₉H₈ClFO₃
MW: 218.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687215
CAS Number: 1323966-33-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR